Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320602.1 | NP_001307531.1 | 2 | 204 | ||
NM_144770.4 | NP_658983.3 | 2 | 204 | |||
NR_135310.1 | 2 | 214 | ||||
NR_135311.1 | 2 | 214 | ||||
XM_005260997.4 | XP_005261054.1 | 2 | 227 | |||
XM_017028385.1 | XP_016883874.1 | 2 | 123 | |||
XM_017028386.1 | XP_016883875.1 | 2 | 359 | |||
XM_017028387.1 | XP_016883876.1 | 2 | 366 | |||
GenBank mRNA | AK091331.1 | - | 2 | 186 | ||
AY077695.1 | - | 2 | 135 | |||
BC030196.1 | - | 2 | 156 | 5199053 | ||
CB153254.1 | - | 2 | 214 | |||
CN430785.1 | - | 2 | 200 |