Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146256.1 | NP_001139728.1 | 3 | 771 | ||
NM_001146257.1 | NP_001139729.1 | 3 | 771 | |||
NM_144969.2 | NP_659406.1 | 4 | 798 | |||
XM_006724624.3 | XP_006724687.1 | 4 | 335 | |||
XM_017029296.1 | XP_016884785.1 | 4 | 335 | |||
GenBank mRNA | AK056374.1 | - | 4 | 336 | ||
AK122885.1 | - | 3 | 520 | |||
AK128587.1 | - | 3 | 306 | |||
AK303896.1 | - | 4 | 336 | |||
AY358786.1 | - | 3 | 692 | |||
BC103980.1 | - | 4 | 336 | 40016292 | ||
BC103981.1 | - | 4 | 336 | 40016293 | ||
BC103982.1 | - | 4 | 336 | 40016294 |