Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144970.2 | NP_659407.1 | 2 | 289 | ||
XM_005272589.2 | XP_005272646.2 | 2 | 289 | |||
XM_006724527.3 | XP_006724590.2 | 2 | 289 | |||
XM_017029302.1 | XP_016884791.1 | 2 | 400 | |||
XM_017029304.1 | XP_016884793.1 | 2 | 259 | |||
XM_017029305.1 | XP_016884794.1 | 2 | 280 | |||
GenBank mRNA | AK095710.1 | - | 1 | 439 | ||
AK123989.1 | - | 2 | 137 | |||
AK315384.1 | - | 2 | 267 | |||
AL832829.1 | - | 2 | 243 | |||
BC025334.1 | - | 2 | 247 | 4559938 | ||
CV572553.1 | - | 2 | 262 | |||
DB207233.1 | - | 2 | 289 |