Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144980.3 | NP_659417.2 | 2 | 144 | ||
XM_005266838.2 | XP_005266895.1 | 2 | 169 | |||
XM_011535509.2 | XP_011533811.1 | 2 | 331 | |||
XM_011535510.2 | XP_011533812.1 | 2 | 331 | |||
XM_011535511.2 | XP_011533813.1 | 2 | 331 | |||
XM_011535512.2 | XP_011533814.1 | 2 | 194 | |||
XM_011535513.2 | XP_011533815.1 | 2 | 597 | |||
XM_017010323.1 | XP_016865812.1 | 2 | 331 | |||
XM_017010324.1 | XP_016865813.1 | 2 | 806 | |||
GenBank mRNA | AK302358.1 | - | 2 | 378 | ||
BC026278.1 | - | 2 | 131 | 4703212 | ||
DB454755.1 | - | 2 | 144 |