Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144683.3 | NP_653284.2 | 3 | 399 | ||
GenBank mRNA | AY358566.1 | - | 3 | 399 | ||
BC015582.2 | - | 2 | 389 | 4637504 |