Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT10 SIRNA Assays ›
Gene Symbol:
SEPT10
Gene Name:
septin 10
Gene Aliases:
-
Chromosome Location:
Chr.2: 109542797 - 109614247 on Build GRCh38
UniGene ID:
Hs.469615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37634
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321496.1
NP_001308425.1
9
1823
NM_001321498.1
NP_001308427.1
12
2220
NM_001321499.1
NP_001308428.1
7
3361
NM_001321500.1
NP_001308429.1
9
3793
NM_001321501.1
NP_001308430.1
10
3866
NM_001321502.1
NP_001308431.1
8
3660
NM_001321503.1
NP_001308432.1
5
3159
NM_001321504.1
NP_001308433.1
7
3479
NM_001321505.1
NP_001308434.1
8
1754
NM_001321506.1
NP_001308435.1
7
1567
NM_001321507.1
NP_001308436.1
8
3548
NM_001321508.1
NP_001308437.1
6
3346
NM_001321509.1
NP_001308438.1
11
2141
NM_001321510.1
NP_001308439.1
8
1636
NM_001321511.1
NP_001308440.1
6
3292
NM_001321512.1
NP_001308441.1
10
3862
NM_001321513.1
NP_001308442.1
10
3999
NM_001321514.1
NP_001308443.1
11
1965
NM_001321515.1
NP_001308444.1
10
2205
NM_144710.4
NP_653311.1
11
2137
NM_178584.3
NP_848699.1
10
2068
XM_011510698.2
XP_011509000.1
12
2053
XM_011510699.2
XP_011509001.1
11
1872
XM_011510700.2
XP_011509002.1
12
2223
XM_011510701.2
XP_011509003.1
10
1947
XM_011510702.2
XP_011509004.1
9
1718
XM_011510703.2
XP_011509005.1
12
1830
XM_011510704.2
XP_011509006.1
9
3729
GenBank mRNA
AB208875.1
-
6
2956
AF146760.1
-
11
1886
AK022682.1
-
8
1469
AK092033.1
-
10
1788
AK125706.1
-
1
1597
AL049934.1
-
5
939
AL353938.1
-
6
1099
BC020502.1
-
11
1831
3913356
BC050345.1
-
12
2158
4830029
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
cell division
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline