Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317791.1 | NP_001304720.1 | 4 | 479 | ||
NM_144721.5 | NP_653322.1 | 5 | 605 | |||
XM_005262772.2 | XP_005262829.1 | 4 | 500 | |||
XM_011531666.2 | XP_011529968.1 | 5 | 1042 | |||
XM_011531667.2 | XP_011529969.1 | 5 | 565 | |||
XM_017007800.1 | XP_016863289.1 | 4 | 645 | |||
GenBank mRNA | AK313765.1 | - | 5 | 989 | ||
AL833350.1 | - | 5 | 565 | |||
BC022989.2 | - | 4 | 519 | 5113206 | ||
BX647579.1 | - | 4 | 439 |