Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 3, 4 | 472 | ||
NM_001300858.1 | NP_001287787.1 | 4, 5 | 683 | |||
NM_144977.4 | NP_659414.2 | 3, 4 | 472 | |||
NR_125340.1 | 3, 4 | 472 | ||||
XM_005244931.2 | XP_005244988.1 | 3, 4 | 472 | |||
XM_006711192.2 | XP_006711255.1 | 4, 5 | 382 | |||
XM_006711193.3 | XP_006711256.1 | 4, 5 | 514 | |||
XM_011509246.2 | XP_011507548.1 | 3, 4 | 333 | |||
XM_011509247.1 | XP_011507549.1 | 3, 4 | 333 | |||
XM_011509248.2 | XP_011507550.1 | 3, 4 | 266 | |||
XM_011509249.2 | XP_011507551.1 | 4, 5 | 571 | |||
XM_011509251.2 | XP_011507553.1 | 3, 4 | 335 | |||
XM_017000470.1 | XP_016855959.1 | 4, 5 | 460 | |||
XM_017000471.1 | XP_016855960.1 | 4, 5 | 382 | |||
XM_017000472.1 | XP_016855961.1 | 1, 2 | 52 | |||
GenBank mRNA | AK091207.1 | - | 3, 4 | 286 | ||
BC022561.1 | - | 4, 5 | 385 | 4793412 | ||
BC063877.1 | - | 3, 4 | 304 | 30377843 | ||
DA495773.1 | - | 3, 4 | 472 |