Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277171.1 | NP_001264100.1 | 3 | 362 | ||
NM_001277173.1 | NP_001264102.1 | 3 | 365 | |||
NM_144979.4 | NP_659416.1 | 3 | 362 | |||
XM_006714119.2 | XP_006714182.1 | 3 | 359 | |||
XM_006714120.1 | XP_006714183.1 | 3 | 359 | |||
XM_011531692.1 | XP_011529994.1 | 3 | 313 | |||
XM_011531693.1 | XP_011529995.1 | 3 | 365 | |||
XM_011531694.1 | XP_011529996.1 | 3 | 216 | |||
XM_011531695.2 | XP_011529997.1 | 3 | 261 | |||
XM_011531696.1 | XP_011529998.1 | 3 | 216 | |||
XM_011531697.2 | XP_011529999.1 | 3 | 359 | |||
GenBank mRNA | AK125877.1 | - | 3 | 365 | ||
AK302720.1 | - | 3 | 299 | |||
BC028588.2 | - | 3 | 286 | 4830259 | ||
DB088314.1 | - | 3 | 361 |