Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304274.1 | NP_001291203.1 | 5 | 496 | ||
NM_144981.2 | NP_659418.1 | 6 | 559 | |||
XM_005252812.3 | XP_005252869.1 | 7 | 680 | |||
XM_011519942.1 | XP_011518244.1 | 7 | 617 | |||
XM_011519943.2 | XP_011518245.1 | 8 | 738 | |||
XM_011519944.2 | XP_011518246.1 | 7 | 646 | |||
XM_011519945.2 | XP_011518247.1 | 6 | 561 | |||
XM_011519946.2 | XP_011518248.1 | 7 | 606 | |||
XM_011519947.2 | XP_011518249.1 | 6 | 543 | |||
XM_017017305.1 | XP_016872794.1 | 6 | 617 | |||
XM_017017306.1 | XP_016872795.1 | 5 | 459 | |||
GenBank mRNA | AK057788.1 | - | 6 | 553 | ||
BC023595.2 | - | 5 | 442 | 4841844 |