Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFC3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144982.4 | NP_659419.3 | 2 | 1099 | ||
GenBank mRNA | AB011118.2 | - | 2 | 949 | ||
BC015679.2 | - | 2 | 988 | |||
BC064336.1 | - | 2 | 980 | |||
BC073843.1 | - | 2 | 954 | |||
BX647709.1 | - | 2 | 979 |