Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPEAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272037.1 | NP_001258966.1 | 3 | 360 | ||
NM_144991.2 | NP_659428.2 | 2 | 298 | |||
GenBank mRNA | AJ487962.1 | - | 2 | 298 | ||
BC021197.2 | - | 2 | 187 | |||
BC115415.1 | - | 2 | 265 | 40080529 |