Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145008.2 | NP_659445.1 | 5 | 1078 | ||
XM_017017318.1 | XP_016872807.1 | 5 | 3284 | |||
GenBank mRNA | AB098738.1 | - | 5 | 1078 | ||
AK054775.1 | - | 5 | 1078 | |||
AK054967.1 | - | 4 | 1867 | |||
AK124577.1 | - | 5 | 3357 | |||
BC029228.1 | - | 4 | 1254 | |||
BC064995.1 | - | 4 | 994 | |||
BC104781.1 | - | 5 | 933 | 8143784 | ||
BC111012.1 | - | 4 | 1268 | |||
BC112119.1 | - | 5 | 933 | 8327587 |