Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP36L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006887.4 | NP_008818.3 | 2 | 1811 | ||
GenBank mRNA | BC005010.1 | - | 2 | 1609 | 3640393 | |
X78992.1 | - | 2 | 1580 |