Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145303.2 | NP_001138775.2 | 10 | 1587 | ||
NM_144686.3 | NP_653287.2 | 10 | 1569 | |||
XM_011526486.2 | XP_011524788.1 | 7 | 1072 | |||
GenBank mRNA | AK124406.1 | - | 2 | 479 | ||
AK314973.1 | - | 10 | 1490 | |||
AY236492.1 | - | 10 | 1586 | |||
AY263166.1 | - | 10 | 1466 | |||
BC025323.1 | - | 10 | 1476 | 2959382 | ||
BC032477.1 | - | 4 | 677 |