Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526528.1 | XP_011524830.1 | 11 | 2752 | ||
XM_011526529.2 | XP_011524831.1 | 11 | 2763 | |||
XM_011526530.1 | XP_011524832.1 | 11 | 2731 | |||
XM_011526531.1 | XP_011524833.1 | 11 | 2719 | |||
XM_011526532.1 | XP_011524834.1 | 11 | 2586 | |||
XM_011526533.2 | XP_011524835.1 | 11 | 2947 | |||
XM_011526534.1 | XP_011524836.1 | 12 | 2752 | |||
XM_011526535.2 | XP_011524837.1 | 9 | 2459 | |||
GenBank mRNA | AK055219.1 | - | 10 | 2973 |