Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144703.2 | NP_653304.2 | 9 | 1662 | ||
XM_005260302.3 | XP_005260359.1 | 10 | 1774 | |||
XM_011528605.2 | XP_011526907.1 | 11 | 1966 | |||
XM_011528606.2 | XP_011526908.1 | 11 | 1884 | |||
XM_011528607.2 | XP_011526909.1 | 10 | 1892 | |||
XM_011528608.2 | XP_011526910.1 | 10 | 1855 | |||
XM_011528609.2 | XP_011526911.1 | 9 | 1736 | |||
XM_011528611.2 | XP_011526913.1 | 8 | 1580 | |||
XM_011528612.2 | XP_011526914.1 | 11 | 1636 | |||
XM_011528613.2 | XP_011526915.1 | 10 | 1496 | |||
XM_011528614.2 | XP_011526916.1 | 10 | 1709 | |||
XM_011528615.2 | XP_011526917.1 | 7 | 1193 | |||
XM_017027688.1 | XP_016883177.1 | 8 | 1569 | |||
XM_017027689.1 | XP_016883178.1 | 8 | 1488 | |||
XM_017027690.1 | XP_016883179.1 | 8 | 1310 | |||
GenBank mRNA | AK058202.1 | - | 9 | 1184 | ||
AK298748.1 | - | 9 | 1371 |