Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320602.1 | NP_001307531.1 | 5 | 1363 | ||
NM_144770.4 | NP_658983.3 | 5 | 1342 | |||
NR_135308.1 | 4 | 1179 | ||||
NR_135309.1 | 4 | 1200 | ||||
NR_135310.1 | 5 | 1352 | ||||
NR_135311.1 | 5 | 1373 | ||||
XM_005260997.4 | XP_005261054.1 | 6 | 1260 | |||
XM_017028384.1 | XP_016883873.1 | 4 | 1101 | |||
XM_017028385.1 | XP_016883874.1 | 4 | 4529 | |||
XM_017028386.1 | XP_016883875.1 | 5 | 1497 | |||
XM_017028387.1 | XP_016883876.1 | 5 | 1504 | |||
GenBank mRNA | AF519623.1 | - | 4 | 1143 | ||
BC030196.1 | - | 5 | 1294 | 5199053 | ||
BQ186146.1 | - | 1 | 41 |