Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308339.1 | NP_001295268.1 | 6, 7 | 1356 | ||
NM_144973.3 | NP_659410.3 | 4, 5 | 1286 | |||
XM_005253326.2 | XP_005253383.1 | 5, 6 | 1288 | |||
XM_005253327.3 | XP_005253384.1 | 4, 5 | 1281 | |||
XM_005253330.3 | XP_005253387.1 | 5, 6 | 757 | |||
XM_006719040.3 | XP_006719103.1 | 5, 6 | 1288 | |||
XM_011520590.2 | XP_011518892.1 | 2, 3 | 524 | |||
XM_017018896.1 | XP_016874385.1 | 5, 6 | 1288 | |||
XM_017018897.1 | XP_016874386.1 | 5, 6 | 1651 | |||
XM_017018899.1 | XP_016874388.1 | 5, 6 | 647 | |||
GenBank mRNA | AK095598.1 | - | 1, 2 | 97 | ||
AK125323.1 | - | 6, 7 | 1356 | |||
BC020855.1 | - | 3, 4 | 956 | |||
BC068580.1 | - | 5, 6 | 1343 | 5265807 |