Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300858.1 | NP_001287787.1 | 16 | 1737 | ||
NM_144977.4 | NP_659414.2 | 16 | 1586 | |||
GenBank mRNA | BC022561.1 | - | 16 | 1439 | 4793412 | |
BC063877.1 | - | 16 | 1418 | 30377843 | ||
H19092.1 | - | 3 | 298 |