Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144980.3 | NP_659417.2 | 3, 4 | 920 | ||
XM_005266838.2 | XP_005266895.1 | 3, 4 | 945 | |||
XM_011535509.2 | XP_011533811.1 | 3, 4 | 1107 | |||
XM_011535511.2 | XP_011533813.1 | 3, 4 | 1107 | |||
XM_011535512.2 | XP_011533814.1 | 3, 4 | 970 | |||
XM_011535513.2 | XP_011533815.1 | 3, 4 | 1373 | |||
XM_017010323.1 | XP_016865812.1 | 3, 4 | 1107 | |||
XM_017010324.1 | XP_016865813.1 | 3, 4 | 1582 | |||
GenBank mRNA | AI333765.1 | - | 2, 3 | 147 | ||
AK302358.1 | - | 3, 4 | 1154 | |||
BC026278.1 | - | 3, 4 | 907 | 4703212 |