Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022478.3 | NP_071923.2 | 3 | 563 | ||
NM_144985.3 | NP_659422.2 | 3 | 563 | |||
XM_011537089.1 | XP_011535391.1 | 3 | 644 | |||
GenBank mRNA | AK057922.1 | - | 3 | 499 | ||
AK289576.1 | - | 3 | 563 | |||
AK307746.1 | - | 3 | 545 | |||
AY260900.1 | - | 2 | 396 | |||
AY260901.1 | - | 2 | 396 | |||
AY358199.1 | - | 3 | 519 | |||
BC110867.1 | - | 3 | 534 | |||
BX248750.1 | - | 3 | 545 |