Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144992.4 | NP_659429.4 | 14 | 2232 | ||
XM_005263897.2 | XP_005263954.1 | 14 | 2197 | |||
XM_006712357.2 | XP_006712420.1 | 13 | 1946 | |||
XM_006712359.3 | XP_006712422.1 | 5 | 626 | |||
XM_006712360.3 | XP_006712423.1 | 2 | 273 | |||
XM_011510770.1 | XP_011509072.1 | 14 | 2197 | |||
XM_011510771.2 | XP_011509073.1 | 13 | 2071 | |||
XM_011510772.1 | XP_011509074.1 | 14 | 2197 | |||
XM_011510774.1 | XP_011509076.1 | 14 | 2197 | |||
XM_017003560.1 | XP_016859049.1 | 8 | 1063 | |||
XM_017003561.1 | XP_016859050.1 | 8 | 1069 | |||
XM_017003562.1 | XP_016859051.1 | 8 | 1119 | |||
XM_017003563.1 | XP_016859052.1 | 8 | 1069 | |||
XM_017003564.1 | XP_016859053.1 | 14 | 2197 | |||
GenBank mRNA | AK057248.1 | - | 11 | 1733 | ||
AK090786.1 | - | 2 | 199 | |||
AK093084.1 | - | 2 | 199 | |||
AK295743.1 | - | 8 | 1127 | |||
AL832635.2 | - | 15 | 2175 | |||
AL832761.1 | - | 14 | 2253 | |||
AL834173.1 | - | 5 | 631 | |||
AL834385.1 | - | 4 | 495 | |||
BC026091.2 | - | 14 | 2047 | |||
BC095480.1 | - | 15 | 2408 | |||
BC150653.1 | - | 14 | 2172 | 9057024 |