Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145016.3 | NP_659453.3 | 3, 4 | 325 | ||
XM_011544809.2 | XP_011543111.1 | 3, 4 | 510 | |||
XM_017017337.1 | XP_016872826.1 | 4, 5 | 460 | |||
XM_017017338.1 | XP_016872827.1 | 3, 4 | 907 | |||
GenBank mRNA | AB207211.1 | - | 2, 3 | 194 | ||
AF426250.1 | - | 3, 4 | 585 | |||
BC016789.1 | - | 4, 5 | 469 | 4091916 | ||
BC021682.1 | - | 3, 4 | 312 | 4079236 |