Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145046.4 | NP_659483.2 | 1, 2 | 146 | ||
GenBank mRNA | AB576176.1 | - | 1, 2 | 83 | ||
AK058084.1 | - | 1, 2 | 138 | |||
BC014595.2 | - | 1, 2 | 146 | 4822010 | ||
BP370084.1 | - | 1, 2 | 126 | |||
DB459545.1 | - | 1, 2 | 148 |