Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145054.4 | NP_659491.4 | 2 | 185 | ||
GenBank mRNA | AB065281.1 | - | 2 | 150 | ||
AK094847.1 | - | 3 | 332 | |||
AK315679.1 | - | 2 | 140 | |||
BC025392.2 | - | 2 | 145 | 4838276 | ||
DB040461.1 | - | 2 | 185 |