Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258205.1 | NP_001245134.1 | 4 | 600 | ||
NM_145068.3 | NP_659505.1 | 4 | 600 | |||
GenBank mRNA | AF514998.1 | - | 4 | 342 | ||
AJ487035.2 | - | 4 | 600 | |||
AY118267.1 | - | 4 | 410 | |||
AY118268.1 | - | 4 | 410 | |||
EU295940.1 | - | 3 | 278 |