Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM200A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145111.3 | NP_659802.1 | 2 | 475 | ||
XM_011515911.2 | XP_011514213.1 | 2 | 279 | |||
XM_011515913.2 | XP_011514215.1 | 2 | 218 | |||
GenBank mRNA | AK094113.1 | - | 2 | 387 | ||
AK290724.1 | - | 2 | 475 | |||
AK315452.1 | - | 2 | 411 | |||
AL713672.1 | - | 1 | 188 | |||
BC009536.1 | - | 2 | 252 | |||
BC040214.1 | - | 2 | 392 | 5166971 | ||
BC126337.1 | - | 2 | 337 | 8992053 | ||
BC130371.1 | - | 2 | 337 | 40146402 | ||
CV569355.1 | - | 2 | 434 |