Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284266.1 | NP_001271195.1 | 2 | 231 | ||
NM_001284267.1 | NP_001271196.1 | 2 | 84 | |||
NM_001284268.1 | NP_001271197.1 | 2 | 242 | |||
NM_001284269.1 | NP_001271198.1 | 2 | 242 | |||
NM_145231.3 | NP_660274.1 | 2 | 231 | |||
GenBank mRNA | AK094740.1 | - | 2 | 184 | ||
AK298930.1 | - | 2 | 176 | |||
AL832321.1 | - | 2 | 168 | |||
BC001787.1 | - | 2 | 143 | 3353520 | ||
BG494945.1 | - | 2 | 196 | |||
BP371056.1 | - | 2 | 84 | |||
BX161414.1 | - | 2 | 208 | |||
BX247991.1 | - | 2 | 153 | |||
CF593330.1 | - | 2 | 227 |