Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145235.3 | NP_660278.3 | 2 | 227 | ||
XM_006718058.3 | XP_006718121.1 | 3 | 208 | |||
XM_006718064.3 | XP_006718127.1 | 2 | 202 | |||
XM_011540348.2 | XP_011538650.1 | 3 | 263 | |||
XM_011540349.2 | XP_011538651.1 | 3 | 267 | |||
XM_011540350.2 | XP_011538652.1 | 2 | 226 | |||
XM_011540351.2 | XP_011538653.1 | 2 | 202 | |||
XM_011540356.2 | XP_011538658.1 | 2 | 192 | |||
XM_011540357.2 | XP_011538659.1 | 3 | 210 | |||
XM_011540358.2 | XP_011538660.1 | 2 | 207 | |||
XM_011540363.2 | XP_011538665.1 | 4 | 426 | |||
XM_017016916.1 | XP_016872405.1 | 3 | 209 | |||
XM_017016917.1 | XP_016872406.1 | 3 | 344 | |||
XM_017016918.1 | XP_016872407.1 | 2 | 151 | |||
XM_017016919.1 | XP_016872408.1 | 2 | 303 | |||
XM_017016920.1 | XP_016872409.1 | 4 | 426 | |||
XM_017016921.1 | XP_016872410.1 | 3 | 344 | |||
XM_017016923.1 | XP_016872412.1 | 2 | 226 | |||
XM_017016924.1 | XP_016872413.1 | 4 | 334 | |||
XM_017016925.1 | XP_016872414.1 | 2 | 221 | |||
GenBank mRNA | AK304407.1 | - | 3 | 278 | ||
AY251163.1 | - | 1 | 127 | |||
AY358154.1 | - | 2 | 205 | |||
BC024189.2 | - | 2 | 227 | 4826296 | ||
BQ225468.1 | - | 2 | 171 | |||
CR627249.1 | - | 2 | 211 |