Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145016.3 | NP_659453.3 | 4 | 343 | ||
XM_011544809.2 | XP_011543111.1 | 4 | 528 | |||
XM_017017337.1 | XP_016872826.1 | 5 | 478 | |||
XM_017017338.1 | XP_016872827.1 | 4 | 925 | |||
GenBank mRNA | AB207211.1 | - | 3 | 212 | ||
AF426250.1 | - | 4 | 603 | |||
BC016789.1 | - | 5 | 487 | 4091916 | ||
BC021682.1 | - | 4 | 330 | 4079236 |