Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111038.1 | NP_001104508.1 | 11 | 1195 | ||
NM_001287150.1 | NP_001274079.1 | 11 | 1195 | |||
NM_145032.3 | NP_659469.3 | 11 | 1372 | |||
NR_105043.1 | 12 | 1286 | ||||
XM_005250205.3 | XP_005250262.1 | 12 | 1278 | |||
XM_005250207.3 | XP_005250264.1 | 12 | 1278 | |||
XM_005250208.3 | XP_005250265.1 | 12 | 1278 | |||
XM_005250209.2 | XP_005250266.1 | 11 | 1176 | |||
XM_006715898.2 | XP_006715961.1 | 6 | 623 | |||
XM_011515928.2 | XP_011514230.1 | 12 | 1278 | |||
XM_011515929.2 | XP_011514231.1 | 12 | 1278 | |||
XM_011515930.2 | XP_011514232.1 | 11 | 1242 | |||
XM_011515932.2 | XP_011514234.1 | 12 | 1278 | |||
XM_017011850.1 | XP_016867339.1 | 12 | 1222 | |||
XM_017011851.1 | XP_016867340.1 | 12 | 1278 | |||
XM_017011852.1 | XP_016867341.1 | 12 | 1428 | |||
XM_017011853.1 | XP_016867342.1 | 11 | 1228 | |||
GenBank mRNA | AK097537.1 | - | 11 | 1372 | ||
AL713709.1 | - | 2 | 94 | |||
AY359238.1 | - | 11 | 1079 | |||
AY359239.1 | - | 11 | 1079 | |||
BC020572.2 | - | 5 | 449 | |||
BC026121.1 | - | 12 | 1289 | 4826546 | ||
BC031285.1 | - | 11 | 1187 | 5298780 |