Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193646.1 | NP_001180575.1 | 3 | 706 | ||
NM_001290746.1 | NP_001277675.1 | 3 | 677 | |||
NM_012068.5 | NP_036200.2 | 4 | 911 | |||
XM_011526629.2 | XP_011524931.1 | 3 | 613 | |||
GenBank mRNA | AB021663.2 | - | 3 | 648 | ||
AB073613.1 | - | 3 | 644 | |||
AF305687.1 | - | 2 | 379 | |||
AK024402.1 | - | 3 | 648 | |||
AK290592.1 | - | 4 | 850 | |||
BC005174.2 | - | 3 | 635 | 3504701 | ||
BM462249.1 | - | 3 | 547 | |||
BU187153.1 | - | 3 | 631 |