Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145166.3 | NP_660149.2 | 6 | 3755 | ||
GenBank mRNA | AB033016.1 | - | 3 | 1757 | ||
AK127065.1 | - | 7 | 3449 | |||
AL133062.1 | - | 5 | 2449 | |||
BC021855.1 | - | 5 | 2412 | 4943271 |