Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145170.3 | NP_660153.3 | 26 | 3282 | ||
XM_005269489.2 | XP_005269546.1 | 25 | 3152 | |||
XM_006717604.2 | XP_006717667.1 | 27 | 4843 | |||
XM_006717605.2 | XP_006717668.1 | 26 | 3245 | |||
XM_006717609.2 | XP_006717672.1 | 24 | 3059 | |||
XM_006717610.2 | XP_006717673.1 | 25 | 4531 | |||
XM_006717611.3 | XP_006717674.1 | 25 | 3183 | |||
XM_011539212.2 | XP_011537514.1 | 22 | 2814 | |||
XM_017015620.1 | XP_016871109.1 | 27 | 4689 | |||
XM_017015621.1 | XP_016871110.1 | 27 | 4686 | |||
XM_017015622.1 | XP_016871111.1 | 27 | 4686 | |||
XM_017015623.1 | XP_016871112.1 | 27 | 4611 | |||
XM_017015624.1 | XP_016871113.1 | 26 | 4608 | |||
XM_017015625.1 | XP_016871114.1 | 26 | 4599 | |||
XM_017015627.1 | XP_016871116.1 | 26 | 4479 | |||
XM_017015628.1 | XP_016871117.1 | 25 | 3061 | |||
XM_017015629.1 | XP_016871118.1 | 25 | 4284 | |||
XM_017015630.1 | XP_016871119.1 | 26 | 4763 | |||
XM_017015631.1 | XP_016871120.1 | 22 | 2620 | |||
XM_017015632.1 | XP_016871121.1 | 25 | 4658 | |||
GenBank mRNA | AF435958.1 | - | 7 | 806 | ||
AF435959.1 | - | 8 | 896 | |||
AK098631.1 | - | 10 | 1292 | |||
BC032856.2 | - | 26 | 3277 | 5267050 |