Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145173.3 | NP_660156.1 | 2 | 1795 | ||
XM_011527713.2 | XP_011526015.1 | 2 | 1960 | |||
GenBank mRNA | AK124445.1 | - | 1 | 1106 | ||
AK124672.1 | - | 1 | 559 | |||
AL833958.1 | - | 1 | 584 | |||
BC030660.1 | - | 2 | 1774 | 4814337 |