Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRSX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145177.2 | NP_660160.2 | 7 | 874 | ||
GenBank mRNA | AJ293620.1 | - | 7 | 834 | ||
AL137300.1 | - | 3 | 448 | |||
AY358849.1 | - | 7 | 836 | |||
BC019696.2 | - | 7 | 827 | |||
BC032340.1 | - | 7 | 838 | 5192975 | ||
BM794858.1 | - | 4 | 554 |