Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143981.1 | NP_001137453.1 | 5 | 754 | ||
NM_001143982.1 | NP_001137454.1 | 5 | 751 | |||
NM_145234.3 | NP_660277.2 | 5 | 599 | |||
XM_005262221.1 | XP_005262278.1 | 5 | 571 | |||
XM_005262222.3 | XP_005262279.1 | 5 | 568 | |||
XM_005262223.1 | XP_005262280.1 | 5 | 548 | |||
XM_005262224.1 | XP_005262281.1 | 5 | 567 | |||
XM_017029959.1 | XP_016885448.1 | 5 | 571 | |||
GenBank mRNA | AB209548.1 | - | 4 | 344 | ||
AK092245.1 | - | 2 | 181 | |||
AK293106.1 | - | 5 | 501 | |||
AK312270.1 | - | 5 | 512 | |||
AY608914.1 | - | 4 | 428 | |||
BC002909.2 | - | 5 | 516 | 3942645 |