Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134793.1 | NP_001128265.1 | 3 | 1712 | ||
NM_145014.2 | NP_659451.1 | 4 | 1465 | |||
XM_005271430.2 | XP_005271487.1 | 2 | 1662 | |||
XM_006718777.3 | XP_006718840.1 | 2 | 979 | |||
XM_011542659.2 | XP_011540961.1 | 2 | 1140 | |||
XM_017017320.1 | XP_016872809.1 | 3 | 1431 | |||
XM_017017321.1 | XP_016872810.1 | 3 | 1428 | |||
GenBank mRNA | AK057477.1 | - | 4 | 1467 | ||
AK127394.1 | - | 3 | 1715 | |||
BC015047.1 | - | 3 | 1053 | 3909690 |