Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145018.3 | NP_659455.3 | 5 | 537 | ||
XM_011544835.1 | XP_011543137.1 | 4 | 399 | |||
XM_011544836.2 | XP_011543138.1 | 4 | 354 | |||
XM_011544837.1 | XP_011543139.1 | 3 | 249 | |||
GenBank mRNA | AK058145.1 | - | 6 | 748 | ||
AK096157.1 | - | 5 | 502 | |||
AK292399.1 | - | 5 | 496 | |||
BC039268.1 | - | 5 | 505 | 5296465 |