Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302453.1 | NP_001289382.1 | 12, 13 | 1519 | ||
NM_001302454.1 | NP_001289383.1 | 10, 11 | 1633 | |||
NM_145045.4 | NP_659482.3 | 12, 13 | 1813 | |||
GenBank mRNA | AK056363.1 | - | 12, 13 | 1734 | ||
AK294355.1 | - | 14, 15 | 1713 | |||
AK302113.1 | - | 12, 13 | 1520 | |||
AK308072.1 | - | 10, 11 | 1595 | |||
BC014252.2 | - | 11, 12 | 1370 | |||
BC141828.1 | - | 12, 13 | 1745 | 40148425 | ||
BC142637.1 | - | 12, 13 | 1758 | 40147788 | ||
BC144421.1 | - | 10, 11 | 1292 |