Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145046.4 | NP_659483.2 | 9 | 1213 | ||
GenBank mRNA | AK058084.1 | - | 9 | 1205 | ||
BC014595.2 | - | 9 | 1213 | 4822010 |