Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080556.1 | NP_001074025.1 | 4 | 477 | ||
NM_145054.4 | NP_659491.4 | 5 | 681 | |||
XM_017024227.1 | XP_016879716.1 | 4 | 466 | |||
GenBank mRNA | AB065281.1 | - | 5 | 646 | ||
AK074435.1 | - | 4 | 461 | |||
AK094847.1 | - | 6 | 828 | |||
AK315679.1 | - | 5 | 636 | |||
BC025392.2 | - | 5 | 641 | 4838276 |