Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAMBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012342.2 | NP_036474.1 | 2, 3 | 775 | ||
GenBank mRNA | AK223065.1 | - | 2, 3 | 747 | ||
AK290639.1 | - | 2, 3 | 767 | |||
BC019252.1 | - | 2, 3 | 760 | 3503549 | ||
BT006820.1 | - | 2, 3 | 372 | |||
U23070.1 | - | 2, 3 | 744 |