Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM84A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145175.2 | NP_660158.2 | 2 | 2616 | ||
GenBank mRNA | AJ417080.1 | - | 2 | 2570 |