Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145233.3 | NP_660276.2 | 4 | 1283 | ||
NR_037801.1 | 4 | 1279 | ||||
GenBank mRNA | BC007868.2 | - | 4 | 1262 | 4303049 | |
BC101589.1 | - | 3 | 1011 | 8069095 | ||
BC101591.1 | - | 3 | 1011 | 8069097 | ||
BC143563.1 | - | 3 | 1011 | 9052074 |
Gene Symbol: ZNF625-ZNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292455.1 | - | 4 | 1279 |