Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145234.3 | NP_660277.2 | 9, 10 | 1175 | ||
GenBank mRNA | BC002909.2 | - | 9, 10 | 1092 | 3942645 |