Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012425.3 | NP_036557.1 | 5 | 487 | ||
NM_152724.2 | NP_689937.2 | 4 | 375 | |||
XM_005252552.3 | XP_005252609.1 | 5 | 570 | |||
GenBank mRNA | AK292911.1 | - | 4 | 300 | ||
AK312520.1 | - | 5 | 430 | |||
BC005993.1 | - | 5 | 457 | 4050837 | ||
BC008691.1 | - | 4 | 646 | 3872071 | ||
BC015644.2 | - | 5 | 447 | 4647836 | ||
BC107898.1 | - | 4 | 354 | |||
BG400752.1 | - | 4 | 376 | |||
BX647733.1 | - | 4 | 351 | |||
CR536521.1 | - | 4 | 344 | |||
CR541840.1 | - | 4 | 344 |