Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323005.1 | NP_001309934.1 | 2 | 481 | ||
NM_145315.4 | NP_660358.2 | 2 | 481 | |||
XM_005266885.3 | XP_005266942.1 | 3 | 1839 | |||
XM_011535657.2 | XP_011533959.1 | 2 | 364 | |||
XM_011535659.2 | XP_011533961.1 | 2 | 364 | |||
XM_011535660.2 | XP_011533962.1 | 2 | 364 | |||
XM_011535661.2 | XP_011533963.1 | 2 | 364 | |||
XM_017010647.1 | XP_016866136.1 | 3 | 331 | |||
XM_017010649.1 | XP_016866138.1 | 3 | 649 | |||
GenBank mRNA | AF520418.1 | - | 2 | 461 | ||
BC018445.1 | - | 2 | 461 | 3871205 | ||
BP246728.1 | - | 2 | 489 |