Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077262.1 | NP_001070730.1 | 3 | 235 | ||
NM_145345.2 | NP_663320.2 | 4 | 454 | |||
NM_183008.2 | NP_892120.2 | 4 | 360 | |||
GenBank mRNA | AF520989.1 | - | 3 | 163 | ||
AF521017.1 | - | 3 | 164 | |||
AK093258.1 | - | 4 | 304 | |||
AK094198.1 | - | 3 | 167 | |||
AK097368.1 | - | 4 | 552 | |||
AK291503.1 | - | 3 | 159 | |||
AK296179.1 | - | 2 | 61 | |||
AY007163.1 | - | 2 | 45 | |||
BC038106.1 | - | 3 | 172 | 5269682 | ||
BX647774.1 | - | 3 | 168 | |||
BX648412.1 | - | 3 | 168 | |||
DA421971.1 | - | 3 | 167 | |||
DA989799.1 | - | 3 | 268 |